[2-Bromo-4-(methylsulphonyl)phenyl]hydrazine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR7916-1G
OR7916-250MG
APOSOR7916-1GEA
116
EUR
APOSOR7916-1G
APOSOR7916-250MG
[2-Bromo-4-(methylsulphonyl)phenyl]hydrazine 95+%
[2-Bromo-4-(methylsulphonyl)phenyl]hydrazine
Formel:
C₇H₉BrN₂O₂S Molecular Weight: 265,13 g/mol Schmelzpunkt: 150…151 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04037940 CAS-Nummer: 849035-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...