[2-Chloro-5-methyl-4-(methylsulphonyl)phenyl]hydrazine 95+%

Lieferant: Apollo Scientific
Achtung

OR7917-1G OR7917-5G OR7917-25G
APOSOR7917-1GEA 285 EUR
APOSOR7917-1G APOSOR7917-5G APOSOR7917-25G
[2-Chloro-5-methyl-4-(methylsulphonyl)phenyl]hydrazine 95+%
[2-Chloro-5-methyl-4-(methylsulphonyl)phenyl]hydrazine
Formel: C₈H₁₁ClN₂O₂S
Molecular Weight: 234,71 g/mol
Schmelzpunkt: 137…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037956
CAS-Nummer: 849035-85-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR