2-Methoxy(benzylhydrazine) dihydrochloride

Lieferant: Apollo Scientific
Achtung

OR7920-1G OR7920-5G OR7920-250MG OR7920-500MG
APOSOR7920-1GEA 829 EUR
APOSOR7920-1G APOSOR7920-5G APOSOR7920-250MG APOSOR7920-500MG
2-Methoxy(benzylhydrazine) dihydrochloride
2-Methoxy(benzylhydrazine) dihydrochloride
Formel: C₈H₁₄Cl₂N₂O
Molecular Weight: 225,12 g/mol
Schmelzpunkt: 117…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03452713
CAS-Nummer: 784189-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR