1-Acetyl-N-(1-benzylpiperidin-4-yl)-2,3-dihydro-1H-indole-6-amine 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-{6-[(1-Benzylpiperidin-4-yl)amino]indolin-1-yl}ethan-1-one

OR7924-250MG OR7924-1G
APOSOR7924-250MGEA 33.5 EUR
APOSOR7924-250MG APOSOR7924-1G
1-Acetyl-N-(1-benzylpiperidin-4-yl)-2,3-dihydro-1H-indole-6-amine 95+%
1-Acetyl-N-(1-benzylpiperidin-4-yl)-2,3-dihydro-1H-indole-6-amine
Formel: C₂₂H₂₇N₃O
Molecular Weight: 349.48 g/mol
Schmelzpunkt: 169…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01319213
CAS-Nummer: 396682-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR