1-(2-Methyl-5-nitrophenyl)biguanid Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR7932-1G OR7932-2.5G
APOSOR7932-1GEA 174 EUR
APOSOR7932-1G APOSOR7932-2.5G
1-(2-Methyl-5-nitrophenyl)biguanid Hydrochlorid
1-(2-Methyl-5-nitrophenyl)biguanid Hydrochlorid
Formel: C₉H₁₃ClN₆O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1170909-72-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR