6-Chloroimidazo[1,2-b]pyridazine-2-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR7936-1G
OR7936-250MG
OR7936-5G
APOSOR7936-1GEA
43.5
EUR
APOSOR7936-1G
APOSOR7936-250MG
APOSOR7936-5G
6-Chloroimidazo[1,2-b]pyridazine-2-carboxylic acid 95%
6-Chloroimidazo[1,2-b]pyridazine-2-carboxylic acid
Formel:
C₇H₄ClN₃O₂ Schmelzpunkt: 253…255 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04974055 CAS-Nummer: 14714-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...