1-(2,6-Dimethoxybenzyl)piperidin-4-one 95+%

Lieferant: Apollo Scientific
Achtung

OR7940-1G OR7940-250MG
APOSOR7940-1GEA 145 EUR
APOSOR7940-1G APOSOR7940-250MG
1-(2,6-Dimethoxybenzyl)piperidin-4-one 95+%
1-(2,6-Dimethoxybenzyl)piperidin-4-one
Formel: C₁₄H₁₉NO₃
Schmelzpunkt: 86…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01319276
CAS-Nummer: 397244-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR