1-Phenyl-2-pyrimidin-4-ylethanone 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR7948-1G
OR7948-5G
OR7948-10G
APOSOR7948-1GEA
109
EUR
APOSOR7948-1G
APOSOR7948-5G
APOSOR7948-10G
1-Phenyl-2-pyrimidin-4-ylethanone 95+%
1-Phenyl-2-pyrimidin-4-ylethanone
Formel:
C₁₂H₁₀N₂O Molecular Weight: 198,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00559708 CAS-Nummer: 36912-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...