1-Phenyl-2-quinoxalin-2-ylethanone 95+%
Lieferant: Apollo Scientific
OR7949-10G
OR7949-5G
OR7949-1G
APOSOR7949-10GEA
606
EUR
APOSOR7949-10G
APOSOR7949-5G
APOSOR7949-1G
1-Phenyl-2-quinoxalin-2-ylethanone 95+%
1-Phenyl-2-quinoxalin-2-ylethanone
Formel:
C₁₆H₁₂N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00483316 CAS-Nummer: 16310-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...