4-Bromo-1-(bromomethyl)-2-(methylsulphonyl)benzene 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-(methylsulphonyl)benzyl bromide

OR7955-1G OR7955-5G OR7955-10G
APOSOR7955-1GEA 89.9 EUR
APOSOR7955-1G APOSOR7955-5G APOSOR7955-10G
4-Bromo-1-(bromomethyl)-2-(methylsulphonyl)benzene 95+%
4-Bromo-1-(bromomethyl)-2-(methylsulphonyl)benzene
Formel: C₈H₈Br₂O₂S
Molecular Weight: 328.03 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037945
CAS-Nummer: 254887-18-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR