1-(Bromomethyl)-4-(methylsulphonyl)-2-nitrobenzene 95+%

Lieferant: Apollo Scientific
Achtung

OR7960-1G OR7960-5G OR7960-25G
APOSOR7960-1GEA 229 EUR
APOSOR7960-1G APOSOR7960-5G APOSOR7960-25G
1-(Bromomethyl)-4-(methylsulphonyl)-2-nitrobenzene 95+%
1-(Bromomethyl)-4-(methylsulphonyl)-2-nitrobenzene
Formel: C₈H₈BrNO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD04037930
CAS-Nummer: 849035-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR