4,5-Dichloro-2-methylphenyl methyl sulphone 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,5-Dichloro-2-(methylsulphonyl)toluene, 1,2-Dichloro-4-methyl-5-(methylsulphonyl)benzene

OR7978-1G OR7978-5G
APOSOR7978-1GEA 154 EUR
APOSOR7978-1G APOSOR7978-5G
4,5-Dichloro-2-methylphenyl methyl sulphone 95+%
4,5-Dichloro-2-methylphenyl methyl sulphone
Formel: C₈H₈Cl₂O₂S
Schmelzpunkt: 137…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037944
CAS-Nummer: 849035-75-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR