1,2-Dichlor-4-(methylsulfonyl)benzol 95+%
Lieferant: Apollo Scientific
OR7982-1G
OR7982-5G
OR7982-25G
APOSOR7982-1GEA
208
EUR
APOSOR7982-1G
APOSOR7982-5G
APOSOR7982-25G
1,2-Dichlor-4-(methylsulfonyl)benzol 95+%
1,2-Dichloro-4-methylsulphonylbenzene
Formel:
C₇H₆Cl₂O₂S Schmelzpunkt: 109…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025063 CAS-Nummer: 38452-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...