2-Methylsulphonyl-1-(pyridin-3-yl)ethanone 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(2-Methylsulphonylacetyl)pyridine

OR7990-1G OR7990-5G OR7990-25G
APOSOR7990-1GEA 136 EUR
APOSOR7990-1G APOSOR7990-5G APOSOR7990-25G
2-Methylsulphonyl-1-(pyridin-3-yl)ethanone 95+%
2-Methylsulphonyl-1-(pyridin-3-yl)ethanone
Formel: C₈H₉NO₃S
Molecular Weight: 199,23 g/mol
Schmelzpunkt: 115…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01941306
CAS-Nummer: 27302-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR