2-Methylsulphonyl-1-(pyridin-2-yl)ethanone 95+%

Lieferant: Apollo Scientific

OR7991-1G OR7991-5G
APOSOR7991-1GEA 130 EUR
APOSOR7991-1G APOSOR7991-5G
2-Methylsulphonyl-1-(pyridin-2-yl)ethanone 95+%
2-Methylsulphonyl-1-(pyridin-2-yl)ethanone
Formel: C₈H₉NO₃S
Molecular Weight: 199,23 g/mol
Schmelzpunkt: 105…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01941305
CAS-Nummer: 27302-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR