3-Methylsulphonylbenzenesulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-(Methylsulphonyl)benzenesulphonyl Chloride

OR7993-1G OR7993-25G OR7993-5G
APOSOR7993-1GEA 21.9 EUR
APOSOR7993-1G APOSOR7993-25G APOSOR7993-5G
3-Methylsulphonylbenzenesulphonyl chloride 95%
3-Methylsulphonylbenzenesulphonyl chloride
Formel: C₇H₇ClO₄S₂
Molecular Weight: 254,71 g/mol
Schmelzpunkt: 93…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037968
CAS-Nummer: 5335-40-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR