3-(Methylsulfonyl)benzonitril 98%

Lieferant: Apollo Scientific
Achtung

OR7994-1G OR7994-25G OR7994-5G
APOSOR7994-1GEA 24.8 EUR
APOSOR7994-1G APOSOR7994-25G APOSOR7994-5G
3-(Methylsulfonyl)benzonitril 98%
3-(Methylsulfonyl)benzonitril
Formel: C₈H₇NO₂S
Molecular Weight: 181,2100 g/mol
Schmelzpunkt: 101…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037969
CAS-Nummer: 22821-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR