2-Amino-3-thiophencarbonitril 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Aminothiophene-3-carbonitrile
OR8015-10G
OR8015-50G
APOSOR8015-10GEA
37.7
EUR
APOSOR8015-10G
APOSOR8015-50G
2-Amino-3-thiophencarbonitril 97%
2-Amino-3-thiophencarbonitril
Formel:
C₅H₄N₂S Molecular Weight: 124,16 g/mol Schmelzpunkt: 97…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00706298 CAS-Nummer: 4651-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...