1-(1-Benzofuran-2-yl)-2-bromoethan-1-one 90%
Lieferant: Apollo Scientific
|
Gefahr
|
OR8021-1G
OR8021-250MG
OR8021-5G
APOSOR8021-1GEA
72.5
EUR
APOSOR8021-1G
APOSOR8021-250MG
APOSOR8021-5G
1-(1-Benzofuran-2-yl)-2-bromoethan-1-one 90%
1-(1-Benzofuran-2-yl)-2-bromoethan-1-one
Formel:
C₁₀H₇BrO₂ Schmelzpunkt: 86…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00176644 CAS-Nummer: 23489-36-3 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...