4-Biphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[1,1'-Biphenyl]-4-ylboronic acid
OR8027-100G
OR8027-25G
OR8027-5G
APOSOR8027-100GEA
87
EUR
APOSOR8027-100G
APOSOR8027-25G
APOSOR8027-5G
4-Biphenylboronsäure 98%
4-Biphenylboronsäure
Formel:
C₁₂H₁₁BO₂ Molecular Weight: 198,03 g/mol Schmelzpunkt: 232…247 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00093311 CAS-Nummer: 5122-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...