(R)-1-Boc-piperidine-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR8030-1G
OR8030-100G
APOSOR8030-1GEA
14.7
EUR
APOSOR8030-1G
APOSOR8030-100G
(R)-1-Boc-piperidine-2-carboxylic acid
(R)-1-Boc-piperidine-2-carboxylic acid
Formel:
C₁₁H₁₉NO₄ Schmelzpunkt: 116…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00237380 CAS-Nummer: 28697-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...