tert-Butyl-4-carbamothioylpiperidine-1-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Carbamothioylpiperidine, N1-BOC protected, tert-Butyl 4-carbamothioylpiperidine-1-carboxylate

OR8046-1G OR8046-5G
APOSOR8046-1GEA 21.9 EUR
APOSOR8046-1G APOSOR8046-5G
tert-Butyl-4-carbamothioylpiperidine-1-carboxylate 95%
tert-Butyl-4-carbamothioylpiperidine-1-carboxylate
Formel: C₁₁H₂₀N₂O₂S
Molecular Weight: 244,3500 g/mol
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02180954
CAS-Nummer: 214834-18-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR