tert-Butyl-3-amino-3-thioxopropylcarbamate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Aminothiopropanamide, N3-BOC protected

OR8047-1G OR8047-5G OR8047-10G
APOSOR8047-1GEA 178 EUR
APOSOR8047-1G APOSOR8047-5G APOSOR8047-10G
tert-Butyl-3-amino-3-thioxopropylcarbamate 95%
tert-Butyl-3-amino-3-thioxopropylcarbamate
Formel: C₈H₁₆N₂O₂S
Molecular Weight: 204,29 g/mol
Schmelzpunkt: 108…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02180883
CAS-Nummer: 77152-97-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR