4-(Chloromethyl)-7-hydroxy-2H-chromen-2-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Chloromethyl)-7-hydroxycoumarin
OR8056-1G
OR8056-5G
APOSOR8056-1GEA
105
EUR
APOSOR8056-1G
APOSOR8056-5G
4-(Chloromethyl)-7-hydroxy-2H-chromen-2-one
4-(Chloromethyl)-7-hydroxy-2H-chromen-2-one
Formel:
C₁₀H₇ClO₃ Molecular Weight: 210.62 g/mol Schmelzpunkt: 179…184 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00467168 CAS-Nummer: 25392-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...