Oxaprozin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(4,5-Diphenyloxazol-2-yl)propanoic acid, 4,5-Diphenyl-2-oxazolepropionic Acid
OR8088-25G
OR8088-5G
APOSOR8088-25GEA
288
EUR
APOSOR8088-25G
APOSOR8088-5G
Oxaprozin 98%
Oxaprozin
Formel:
C₁₈H₁₅NO₃ Molecular Weight: 293,32 g/mol Schmelzpunkt: 154…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00215977 CAS-Nummer: 21256-18-8 EINECS: 244-296-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...