Methyl-4-amino-2,2'-bithiophene-5-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-amino-5-(thien-2-yl)thiophene-2-carboxylate, Methyl 4-amino-[2,2'-bithiophene]-5-carboxylate

OR8116-1G OR8116-500MG
APOSOR8116-1GEA 306 EUR
APOSOR8116-1G APOSOR8116-500MG
Methyl-4-amino-2,2'-bithiophene-5-carboxylate 97%
Methyl-4-amino-2,2'-bithiophene-5-carboxylate
Formel: C₁₀H₉NO₂S₂
Molecular Weight: 239,32 g/mol
Schmelzpunkt: 107…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01566302
CAS-Nummer: 169759-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR