3-Methylbenzamidin Hydrochlorid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methylbenzoesäureamidin Hydrochlorid, 3-Methylphenylamidin Hydrochlorid
OR8117-1G
OR8117-25G
OR8117-5G
APOSOR8117-1GEA
69.8
EUR
APOSOR8117-1G
APOSOR8117-25G
APOSOR8117-5G
3-Methylbenzamidin Hydrochlorid 98%
3-Methylbenzamidin Hydrochlorid
Formel:
C₈H₁₁ClN₂ Molecular Weight: 170,64 g/mol Schmelzpunkt: 186…188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02180878 CAS-Nummer: 20680-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...