3-(5-Methylfuran-2-yl)phenylamine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(5-Methylfur-2-yl)aniline

OR8126-1G OR8126-250MG
APOSOR8126-1GEA 232 EUR
APOSOR8126-1G APOSOR8126-250MG
3-(5-Methylfuran-2-yl)phenylamine 97%
3-(5-Methylfuran-2-yl)phenylamine
Formel: C₁₁H₁₁NO
Molecular Weight: 173.22 g/mol
Schmelzpunkt: 61…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01126401
CAS-Nummer: 306935-67-1
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR