4-(Piperazin-1-yl)-1H-indole
Lieferant: Apollo Scientific
Synonyme:
4-(1-Piperazinyl)indole
OR8146-1G
OR8146-250MG
APOSOR8146-1GEA
340
EUR
APOSOR8146-1G
APOSOR8146-250MG
4-(Piperazin-1-yl)-1H-indole
4-(Piperazin-1-yl)-1H-indole
Formel:
C₁₂H₁₅N₃ Molecular Weight: 201,27 g/mol Schmelzpunkt: 198…199 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00831776 CAS-Nummer: 84807-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...