N-Acetylneuraminic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Neu5Ac, (4S,5R,6R,7S,8R)-5-Acetamido-4,6,7,8,9-pentahydroxy-2-oxononanoic acid, NANA, Sialic acid, 5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-nonulopyranosonic acid, Lactaminic acid
OR8154-5G
OR8154-10G
APOSOR8154-5GEA
33.5
EUR
APOSOR8154-5G
APOSOR8154-10G
N-Acetylneuraminic acid 97%
N-Acetylneuraminic acid
Formel:
C₁₁H₁₉NO₉ Molecular Weight: 309,27 g/mol Schmelzpunkt: 184…186 °C (Zers.) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00006620 CAS-Nummer: 131-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...