Methyl-2-amino-3-nitrobenzoat 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-amino-3-nitrobenzoate
OR8159-1G
OR8159-25G
OR8159-5G
APOSOR8159-1GEA
21.9
EUR
APOSOR8159-1G
APOSOR8159-25G
APOSOR8159-5G
Methyl-2-amino-3-nitrobenzoat 95%
Methyl-2-amino-3-nitrobenzoat
Formel:
C₈H₈N₂O₄ Molecular Weight: 196,1600 g/mol Schmelzpunkt: 97…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093531 CAS-Nummer: 57113-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...