5-Nitro-2-(morpholin-4-yl)benzoic acid
Lieferant: Apollo Scientific
Synonyme:
2-Morpholin-4-yl-5-nitrobenzoic acid, 2-Morpholino-5-nitrobenzoic acid
OR8190-10G
OR8190-1G
OR8190-5G
APOSOR8190-10GEA
419
EUR
APOSOR8190-10G
APOSOR8190-1G
APOSOR8190-5G
5-Nitro-2-(morpholin-4-yl)benzoic acid
5-Nitro-2-(morpholin-4-yl)benzoic acid
Formel:
C₁₁H₁₂N₂O₅ Molecular Weight: 252,23 g/mol Schmelzpunkt: 167…168 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00207115 CAS-Nummer: 4036-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...