5-Nitro-2-(2,6-dimethylmorpholin-4-yl)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2,6-Dimethylmorpholin-4-yl)-5-nitrobenzoic acid
OR8191-1G
OR8191-250MG
APOSOR8191-1GEA
87
EUR
APOSOR8191-1G
APOSOR8191-250MG
5-Nitro-2-(2,6-dimethylmorpholin-4-yl)benzoic acid
5-Nitro-2-(2,6-dimethylmorpholin-4-yl)benzoic acid
Formel:
C₁₃H₁₆N₂O₅ Schmelzpunkt: 172 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052231 CAS-Nummer: 175136-71-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...