3-Iodbenzaldehyd 99%

Lieferant: Apollo Scientific

OR8221-1G OR8221-25G OR8221-5G
APOSOR8221-1GEA 21.9 EUR
APOSOR8221-1G APOSOR8221-25G APOSOR8221-5G
3-Iodbenzaldehyd 99%
3-Iodobenzaldehyde
Formel: C₇H₅IO
Molecular Weight: 232,02 g/mol
Siedepunkt: 124…125 °C/13mmHg
Schmelzpunkt: 55…60 °C
MDL: MFCD00039573
CAS-Nummer: 696-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR