2-Amino-4-(4-methylphenyl)pyrimidine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(p-Tolyl)pyrimidin-2-amine, 4-(4-Methylphenyl)pyrimidin-2-amine

OR8227-1G OR8227-500MG
APOSOR8227-1GEA 46.4 EUR
APOSOR8227-1G APOSOR8227-500MG
2-Amino-4-(4-methylphenyl)pyrimidine
2-Amino-4-(4-methylphenyl)pyrimidine
Formel: C₁₁H₁₁N₃
Molecular Weight: 185,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665911
CAS-Nummer: 263276-44-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR