4-Nitrophenethylbromid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-Nitrophenyl)-1-bromoethane, 1-(2-Bromoethyl)-4-nitrobenzene
OR8248-10G
OR8248-50G
APOSOR8248-10GEA
14.7
EUR
APOSOR8248-10G
APOSOR8248-50G
4-Nitrophenethylbromid
4-Nitrophenethylbromid
Formel:
C₈H₈BrNO₂ Molecular Weight: 230,06 g/mol Siedepunkt: 116 °C/0,2mmHg Schmelzpunkt: 67…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007386 CAS-Nummer: 5339-26-4 EINECS: 226-271-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...