3'-Azido-3'-desoxythymidin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Azido-2,3-dideoxy-beta-D-ribofuranosyl)thymine, Zidovudine, Azidothymidine, AZT

OR8250T-1G OR8250T-5G
APOSOR8250T-1GEA 14.7 EUR
APOSOR8250T-1G APOSOR8250T-5G
3'-Azido-3'-desoxythymidin 97%
3'-Azido-3'-desoxythymidin

A potent & selective inhibitor of HIV-1 replication.

Formel: C₁₀H₁₃N₅O₄
Molecular Weight: 267,25 g/mol
Schmelzpunkt: 113…115 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00006536
CAS-Nummer: 30516-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR