3-Brom-5-nitro-N-propylbenzamid 98%
Lieferant: Apollo Scientific
OR8251-1G
OR8251-5G
APOSOR8251-1GEA
132
EUR
APOSOR8251-1G
APOSOR8251-5G
3-Brom-5-nitro-N-propylbenzamid 98%
3-Brom-5-nitro-N-propylbenzamid
Formel:
C₁₀H₁₁BrN₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258764 CAS-Nummer: 929000-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...