trans-2,4-Dichlorzimtsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dichlorcinnamsäure, 3-(2,4-Dichlorphenyl)propensäure

OR8283-25G OR8283-5G
APOSOR8283-25GEA 30.6 EUR
APOSOR8283-25G APOSOR8283-5G
trans-2,4-Dichlorzimtsäure 98%
trans-2,4-Dichlorzimtsäure
Formel: C₉H₆Cl₂O₂
Molecular Weight: 217.05 g/mol
Schmelzpunkt: 233…235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004373
CAS-Nummer: 20595-45-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR