4-Amino-6-chlorpyrimidin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chlorpyrimidin-4-ylamin

OR8288-25G OR8288-100G OR8288-500G
APOSOR8288-25GEA 21.9 EUR
APOSOR8288-25G APOSOR8288-100G APOSOR8288-500G
4-Amino-6-chlorpyrimidin 97%
4-Amino-6-chloropyrimidine
Formel: C₄H₄ClN₃
Molecular Weight: 129.55 g/mol
Schmelzpunkt: 214…219 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053576
CAS-Nummer: 5305-59-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR