8-Methyl-4-chinazolon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Methylquinazolin-4(3H)-one

OR8297-1G OR8297-250MG
APOSOR8297-1GEA 52.2 EUR
APOSOR8297-1G APOSOR8297-250MG
8-Methyl-4-chinazolon 98%
8-Methyl-4-chinazolon
Formel: C₉H₈N₂O
Molecular Weight: 160,1726 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01685945
CAS-Nummer: 19181-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR