2-Amino-3-brom-5-chlorpyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Brom-5-chlorpyridin-2-amin, 3-Brom-5-chlor-2-pyridylamin
OR8308-1G
OR8308-5G
APOSOR8308-1GEA
14.7
EUR
APOSOR8308-1G
APOSOR8308-5G
2-Amino-3-brom-5-chlorpyridin 98%
2-Amino-3-brom-5-chlorpyridin
Formel:
C₅H₄BrClN₂ Molecular Weight: 207,46 g/mol Schmelzpunkt: 83…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00955627 CAS-Nummer: 26163-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...