7-Brom-5-chlor-1H-indazol 97%

Lieferant: Apollo Scientific
Achtung

OR8313-1G OR8313-5G OR8313-10G
APOSOR8313-1GEA 105 EUR
APOSOR8313-1G APOSOR8313-5G APOSOR8313-10G
7-Brom-5-chlor-1H-indazol 97%
7-Bromo-5-chloro-1H-indazole
Formel: C₇H₄BrClN₂
Lagertemperatur: Kühlschrank
MDL: MFCD09258643
CAS-Nummer: 875305-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR