N-Boc-2-pyrroleboronic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H-Pyrrole-2-boronic acid, N-BOC protected, N-Boc-2-Pyrroleboronic acid
OR8332-5G
OR8332-1G
APOSOR8332-5GEA
49.3
EUR
APOSOR8332-5G
APOSOR8332-1G
N-Boc-2-pyrroleboronic acid 95%
N-Boc-2-pyrroleboronic acid
Formel:
C₉H₁₄BNO₄ Molecular Weight: 211,02 g/mol Schmelzpunkt: 93…98 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD01318939 CAS-Nummer: 135884-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...