2-Chloro-4-formylthiazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-1,3-thiazole-4-carboxaldehyde

OR8404-100MG OR8404-1G OR8404-250MG
APOSOR8404-100MGEA 29 EUR
APOSOR8404-100MG APOSOR8404-1G APOSOR8404-250MG
2-Chloro-4-formylthiazole
2-Chloro-4-formylthiazole
Formel: C₄H₂ClNOS
Molecular Weight: 147,5828 g/mol
Schmelzpunkt: 104…105 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09702023
CAS-Nummer: 5198-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR