2-Chloro-7H-pyrrolo[2,3-d]pyrimidine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR8407-1G
OR8407-5G
APOSOR8407-1GEA
26.1
EUR
APOSOR8407-1G
APOSOR8407-5G
2-Chloro-7H-pyrrolo[2,3-d]pyrimidine 95%
2-Chloro-7H-pyrrolo[2,3-d]pyrimidine
Formel:
C₆H₄ClN₃ Molecular Weight: 153.5700 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07787392 CAS-Nummer: 335654-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...