(Z)-1-Ethoxyethene-2-boronic acid pinacol ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(Z)-2-(2-Ethoxyvinyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
OR8411-1G
OR8411-5G
APOSOR8411-1GEA
319
EUR
APOSOR8411-1G
APOSOR8411-5G
(Z)-1-Ethoxyethene-2-boronic acid pinacol ester
(Z)-1-Ethoxyethene-2-boronic acid pinacol ester
Formel:
C₁₀H₁₉BO₃ Molecular Weight: 198,07 g/mol Siedepunkt: 50 °C/0,3mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD09702441 CAS-Nummer: 219489-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...