5-Hydroxyisatosäureanhydrid

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Hydroxy-1H-benzo[d][1,3]oxazine-2,4-dione

OR8420-10G OR8420-1G OR8420-5G OR8420-25G
APOSOR8420-10GEA 248 EUR
APOSOR8420-10G APOSOR8420-1G APOSOR8420-5G APOSOR8420-25G
5-Hydroxyisatosäureanhydrid
5-Hydroxyisatosäureanhydrid
Formel: C₈H₅NO₄
Molecular Weight: 179,1296 g/mol
Schmelzpunkt: 275…279 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00798201
CAS-Nummer: 195986-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR