1-(2-Nitrophenyl)piperidine-4-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR8450-1G
OR8450-5G
APOSOR8450-1GEA
141
EUR
APOSOR8450-1G
APOSOR8450-5G
1-(2-Nitrophenyl)piperidine-4-carboxylic acid
1-(2-Nitrophenyl)piperidine-4-carboxylic acid
Formel:
C₁₂H₁₄N₂O₄ Schmelzpunkt: 171…172 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06362460 CAS-Nummer: 438192-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...