1-(3-Nitro-2-pyridinyl)piperazin

Lieferant: Apollo Scientific

Synonyme: 1-(3-Nitropyridin-2-yl)piperazine

OR8469-5G
APOSOR8469-5GEA 150 EUR
APOSOR8469-5G
1-(3-Nitro-2-pyridinyl)piperazin
1-(3-Nitro-2-pyridinyl)piperazin
Formel: C₉H₁₂N₄O₂
Molecular Weight: 208.2172 g/mol
Schmelzpunkt: 84…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05182227
CAS-Nummer: 87394-48-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR